Iridium(III) chloride hydrate

Leverandør: TCI
Warning

I0616-1G I0616-5G
TCIAI0616-1GEA 3300 NOK
TCIAI0616-1G TCIAI0616-5G
Iridium(III) chloride hydrate
Iridium(III) chloride hydrate
Formel: Cl₃Ir·xH₂O
Tetthet: 5,3 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149750
CAS-nummer: 14996-61-3
EINECS: 233-044-6
Merck Indeks: 14,05088
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b3edb7aa4ebed506f731814bee81c337129dd81&vid=6b3edb7aa4ebed506f731814bee81c337129dd81&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Dark green to Black powder to crystal
Iridium 50.0 to 56.0 %
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR