(R)-(+)-α-Methylbenzylamine ≥99.0% (by GC)
Leverandør: TCI
Danger
Synonymer: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
P0794-25ML P0794-500ML P0794-100ML
TCIAP0794-25MLEA 405 NOK
TCIAP0794-25ML TCIAP0794-500ML TCIAP0794-100ML
(R)-(+)-α-Methylbenzylamine ≥99.0% (by GC)
(R)-(+)-α-Methylbenzylamine
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokepunkt: 85°C
Smeltepunkt: 32,5 °C
Tetthet: 0,95 g/cm³ (20 °C)
Flash punkt: 70°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064405
CAS-nummer: 3886-69-9
EINECS: 223-423-4
FN: 2922
ADR: 8,II
Merck Indeks: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9074e348f042639b9094ac142701e2f83761b174&vid=9074e348f042639b9094ac142701e2f83761b174&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Specific rotation [a]20/D +37.0 to +41.0 deg (neat)