Ethyl laurate ≥99.0% (by GC)
Leverandør: TCI
Synonymer:
Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester
L0013-25ML
L0013-500ML
TCIAL0013-25MLEA
220
NOK
TCIAL0013-25ML
TCIAL0013-500ML
Ethyl laurate ≥99.0% (by GC)
Ethyl laurate
Formel:
C₁₄H₂₈O₂ MW: 228,38 g/mol Kokepunkt: 275°C Smeltepunkt: -10°C Tetthet: 0,86 g/cm³ (20 °C) Flash punkt: 101°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00015065 CAS-nummer: 106-33-2 EINECS: 203-386-0 Merck Indeks: 14,03818 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Acid Value | max. 0.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...