Ethyl hexanoate ≥99.0% (by GC)
Leverandør: TCI
Warning
Synonymer: Ethyl caproate, Caproic acid ethyl ester, Hexanoic acid ethyl ester
H0108-25ML H0108-500ML
TCIAH0108-25MLEA 220 NOK
TCIAH0108-25ML TCIAH0108-500ML
Ethyl hexanoate ≥99.0% (by GC)
Ethyl hexanoate
Formel: C₈H₁₆O₂
MW: 144,21 g/mol
Kokepunkt: 168 °C (1013 hPa)
Smeltepunkt: –67 °C
Tetthet: 0,87 g/cm³ (20 °C)
Flash punkt: 55°C
MDL nummer: MFCD00009511
CAS-nummer: 123-66-0
EINECS: 204-640-3
FN: 3272
ADR: 3,III
Merck Indeks: 14,03777
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f281167feb22a4941bd121c0fea959b0c862c36&vid=6f281167feb22a4941bd121c0fea959b0c862c36&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %