Sarcosine ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

M0332-25G M0332-500G M0332-100G
TCIAM0332-25GEA 405 NOK
TCIAM0332-25G TCIAM0332-500G TCIAM0332-100G
Sarcosine ≥98.0% (by titrimetric analysis)
Sarcosine
Formel: C₃H₇NO₂
MW: 89,09 g/mol
Kokepunkt: 185 °C (1013 hPa)
Smeltepunkt: 63 °C
Tetthet: 1,301 g/cm³ (20 °C)
MDL nummer: MFCD00004279
CAS-nummer: 107-97-1
EINECS: 203-538-6
Merck Indeks: 14,08373
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29ebb0d7572f52a06b622d3839c45ceb21a210fb&vid=29ebb0d7572f52a06b622d3839c45ceb21a210fb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR