2-Ethylaminoethanol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol

E0057-25ML E0057-500ML E0057-100ML
TCIAE0057-25MLEA 220 NOK
TCIAE0057-25ML TCIAE0057-500ML TCIAE0057-100ML
2-Ethylaminoethanol ≥98.0% (by GC, titration analysis)
2-Ethylaminoethanol
Formel: C₄H₁₁NO
MW: 89,14 g/mol
Kokepunkt: 169°C
Smeltepunkt: –9 °C
Tetthet: 0,914 g/cm³ (20 °C)
Flash punkt: 71°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002841
CAS-nummer: 110-73-6
EINECS: 203-797-5
FN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d2dc9519fd780b6d978402c2f1a855fea2f4880&vid=6d2dc9519fd780b6d978402c2f1a855fea2f4880&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR