1-Butanesulphonyl chloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 1-Butanesulphonylchloride

B0866-25G B0866-500G
TCIAB0866-25GEA 865 NOK
TCIAB0866-25G TCIAB0866-500G
1-Butanesulphonyl chloride ≥98.0% (by titrimetric analysis)
1-Butanesulphonyl chloride
Formel: C₄H₉ClO₂S
MW: 156,63 g/mol
Kokepunkt: 65°C
Smeltepunkt: –25 °C
Tetthet: 1,2404 g/cm³ (0 °C)
Flash punkt: 79°C
MDL nummer: MFCD00007464
CAS-nummer: 2386-60-9
EINECS: 219-204-8
FN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e22c4c8f5c27453d18463c6cff08ecbefebed39&vid=5e22c4c8f5c27453d18463c6cff08ecbefebed39&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR