cis-3-Hexen-1-ol ≥97.0% (by GC)

Leverandør: TCI

Synonymer: cis-Hex-3-en-1-ol

H0124-25ML H0124-100ML
TCIAH0124-25MLEA 650 NOK
TCIAH0124-25ML TCIAH0124-100ML
cis-3-Hexen-1-ol ≥97.0% (by GC)
cis-3-Hexen-1-ol
Formel: C₆H₁₂O
MW: 100,16 g/mol
Kokepunkt: 156 °C (1013 hPa)
Smeltepunkt: –60 °C
Tetthet: 0,85 g/cm³ (20 °C)
Flash punkt: 62°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063217
CAS-nummer: 928-96-1
EINECS: 213-192-8
ADR: 3,III
Merck Indeks: 13,04719
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97f2310e62aa9c67cfb213631123497393f3b37f&vid=97f2310e62aa9c67cfb213631123497393f3b37f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR