trans-2-Hepten-1-ol ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: trans-Hept-2-en-1-ol

H0682-25ML H0682-5ML
TCIAH0682-25MLEA 2575 NOK
TCIAH0682-25ML TCIAH0682-5ML
trans-2-Hepten-1-ol ≥96.0% (by GC)
trans-2-Hepten-1-ol
Formel: C₇H₁₄O
MW: 114,19 g/mol
Kokepunkt: 75°C
Tetthet: 0,8516 g/cm³ (20 °C)
Flash punkt: 57°C
MDL nummer: MFCD00014056
CAS-nummer: 33467-76-4
EINECS: 251-534-8
FN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd5207bd3cca29b66f0f4af6d604943701a4e273&vid=bd5207bd3cca29b66f0f4af6d604943701a4e273&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR