3-Methylpiperidine ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Danger
P0444-25ML P0444-500ML
TCIAP0444-25MLEA 220 NOK
TCIAP0444-25ML TCIAP0444-500ML
3-Methylpiperidine ≥97.0% (by GC, titration analysis)
3-Methylpiperidine
Formel: C₆H₁₃N
MW: 99,18 g/mol
Kokepunkt: 126°C
Smeltepunkt: –24 °C
Tetthet: 0,8635 g/cm³ (0 °C)
Flash punkt: 8°C
MDL nummer: MFCD00005994
CAS-nummer: 626-56-2
EINECS: 210-953-6
FN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cedba0150f6121b49a93e9a08be3f8c22ce57dcd&vid=cedba0150f6121b49a93e9a08be3f8c22ce57dcd&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %