1-Bromo-3,5-dimethoxybenzene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
B3848-25G
B3848-5G
TCIAB3848-25GEA
1050
NOK
TCIAB3848-25G
TCIAB3848-5G
1-Bromo-3,5-dimethoxybenzene ≥98.0% (by GC)
1-Bromo-3,5-dimethoxybenzene
Formel:
C₈H₉BrO₂ MW: 217.06 g/mol |
MDL nummer:
MFCD06200685 CAS-nummer: 20469-65-2 |
|
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...