2-Cyanobenzenesulphonyl chloride ≥97.0% (by GC)
Leverandør: TCI
Danger
Synonymer: 2-Cyanobenzene-1-sulphonyl chloride
C1811-1G C1811-5G
TCIAC1811-1GEA 525 NOK
TCIAC1811-1G TCIAC1811-5G
2-Cyanobenzenesulphonyl chloride ≥97.0% (by GC)
2-Cyanobenzenesulphonyl chloride
Formel: C₇H₄ClNO₂S
MW: 201,63 g/mol
Smeltepunkt: 67,5 °C
MDL nummer: MFCD00052341
CAS-nummer: 69360-26-5
FN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcfd428d463944611a083bf6eed37ccf3f2ae709&vid=dcfd428d463944611a083bf6eed37ccf3f2ae709&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 96.0 %
Melting point 65.0 to 70.0 deg-C