1-Nitro-4-(trifluoromethoxy)benzene ≥96.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
4-(Trifluoromethoxy)nitrobenzene
T2155-25G
T2155-5G
TCIAT2155-25GEA
1025
NOK
TCIAT2155-25G
TCIAT2155-5G
1-Nitro-4-(trifluoromethoxy)benzene ≥96.0% (by GC)
1-Nitro-4-(trifluoromethoxy)benzene
Formel:
C₇H₄F₃NO₃ MW: 207,11 g/mol Kokepunkt: 60°C Smeltepunkt: 15°C |
MDL nummer:
MFCD00040302 CAS-nummer: 713-65-5 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...