3,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 3,4-Dimethoxyphenylisothiocyanate, 1,2-Dimethoxy-4-isothiocyanatobenzene

I0541-25G I0541-5G
TCIAI0541-25GEA 3125 NOK
TCIAI0541-25G TCIAI0541-5G
3,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)
3,4-Dimethoxyphenyl isothiocyanate
Formel: C₉H₉NO₂S
MW: 195,24 g/mol
Kokepunkt: 171 - 172 °C (7 -rr)
Smeltepunkt: 48…48,5 °C
MDL nummer: MFCD00041075
CAS-nummer: 33904-04-0
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5fb4d377c92e6127659d5e4871be1c4b471fbea&vid=b5fb4d377c92e6127659d5e4871be1c4b471fbea&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Butylamine Method) min. 96.0 %
Melting point 46.0 to 50.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR