3-(Chloromethyl)benzoyl chloride ≥93.0% (by GC)

Leverandør: TCI
Danger

C2314-25G C2314-5G
TCIAC2314-25GEA 2050 NOK
TCIAC2314-25G TCIAC2314-5G
3-(Chloromethyl)benzoyl chloride ≥93.0% (by GC)
3-(Chloromethyl)benzoyl chloride
Formel: C₈H₆Cl₂O
MW: 189,04 g/mol
Kokepunkt: 150°C
Smeltepunkt: 4°C
Tetthet: 1,331 g/cm³ (20 °C)
MDL nummer: MFCD00000682
CAS-nummer: 63024-77-1
EINECS: 263-799-7
FN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b4ceea48e98357c43752679895478b4e458a624&vid=0b4ceea48e98357c43752679895478b4e458a624&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR