3-(Trifluoromethoxy)benzyl alcohol ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
T1955-1G
T1955-5G
TCIAT1955-1GEA
540
NOK
TCIAT1955-1G
TCIAT1955-5G
3-(Trifluoromethoxy)benzyl alcohol ≥98.0% (by GC)
3-(Trifluoromethoxy)benzyl alcohol
Formel:
C₈H₇F₃O₂ MW: 192,14 g/mol |
MDL nummer:
MFCD00061270 CAS-nummer: 50823-90-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...