2'-Iodacetofenon ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-(2-Iodophenyl)ethan-1-one, 1-(2-Iodophenyl)ethanone, o-Iodoacetophenone

I0689-1G I0689-5G
TCIAI0689-1GEA 325 NOK
TCIAI0689-1G TCIAI0689-5G
2'-Iodacetofenon ≥98.0% (by GC)
2'-Iodacetofenon
Formel: C₈H₇IO
MW: 246,05 g/mol
Kokepunkt: 144°C
Flash punkt: 103°C
MDL nummer: MFCD00094998
CAS-nummer: 2142-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de8d0dd49422da331ae2fc3cf87ef27f5f290e04&vid=de8d0dd49422da331ae2fc3cf87ef27f5f290e04&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR