3,5-Dimethoxybenzyl alcohol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (3,5-Dimethoxyphenyl)methanol

D2594-25G D2594-5G
TCIAD2594-25GEA 2100 NOK
TCIAD2594-25G TCIAD2594-5G
3,5-Dimethoxybenzyl alcohol ≥98.0% (by GC)
3,5-Dimethoxybenzyl alcohol
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
Kokepunkt: 171°C
Smeltepunkt: 48…50 °C
MDL nummer: MFCD00004641
CAS-nummer: 705-76-0
EINECS: 211-888-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98795859116a5fbd8445ed82600108f3fc5724bd&vid=98795859116a5fbd8445ed82600108f3fc5724bd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 51.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR