3-fenoksybensyl alkohol
Leverandør: TCI
|
Warning
|
Synonymer:
3-(Hydroxymethyl)diphenyl ether, (3-Phenoxyphenyl)methanol
P0979-25G
P0979-500G
TCIAP0979-25GEA
475
NOK
TCIAP0979-25G
TCIAP0979-500G
3-fenoksybensyl alkohol
3-fenoksybensyl alkohol
Formel:
C₁₃H₁₂O₂ MW: 200,24 g/mol Kokepunkt: 300°C Tetthet: 1,146 g/cm³ (20 °C) |
MDL nummer:
MFCD00004636 CAS-nummer: 13826-35-2 EINECS: 237-525-1 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...