Ethyl toluene-4-sulphonate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester
T0268-25G
T0268-500G
TCIAT0268-25GEA
220
NOK
TCIAT0268-25G
TCIAT0268-500G
Ethyl toluene-4-sulphonate ≥98.0% (by GC)
Ethyl toluene-4-sulphonate
Formel:
C₉H₁₂O₃S MW: 200,26 g/mol Kokepunkt: 173°C Smeltepunkt: 30…32 °C Tetthet: 1,174 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00009100 CAS-nummer: 80-40-0 EINECS: 201-276-7 ADR: 6.1,III Merck Indeks: 14,03858 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Water | max. 0.5 % |
Acid | max. 0.3 % (as p-Toluenesulfonic Acid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...