1-Pentyl-4-ethynylbenzene ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 4-N-Pentylphenylacetylene, 4-n-Pentylphenylacetylene, (4-Pentylphenyl)acetylene, 1-Ethynyl-4-pentylbenzene, (4-Pentylphenyl)ethyne

E0563-25G E0563-5G
TCIAE0563-25GEA 3625 NOK
TCIAE0563-25G TCIAE0563-5G
1-Pentyl-4-ethynylbenzene ≥97.0% (by GC)
1-Pentyl-4-ethynylbenzene
Formel: C₁₃H₁₆
MW: 172,27 g/mol
Flash punkt: 104°C
MDL nummer: MFCD00173884
CAS-nummer: 79887-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1591f690f633967aec13a0977370236a6167daed&vid=1591f690f633967aec13a0977370236a6167daed&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR