2,4,6-Trimethylbenzyl alcohol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (2,4,6-Trimethylphenyl)methanol, Mesitylmethanol

T1905-25G T1905-5G
TCIAT1905-25GEA 3825 NOK
TCIAT1905-25G TCIAT1905-5G
2,4,6-Trimethylbenzyl alcohol ≥98.0% (by GC)
2,4,6-Trimethylbenzyl alcohol
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokepunkt: 141°C
Smeltepunkt: 88 °C
MDL nummer: MFCD00014422
CAS-nummer: 4170-90-5
EINECS: 224-032-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=412b933f0a22620cac643469faf0f4c1715266b4&vid=412b933f0a22620cac643469faf0f4c1715266b4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 90.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR