2-Nitrobifenyl ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
N0194-25G
N0194-500G
TCIAN0194-25GEA
620
NOK
TCIAN0194-25G
TCIAN0194-500G
2-Nitrobifenyl ≥98.0% (by GC)
2-Nitrobifenyl
Formel:
C₁₂H₉NO₂ MW: 199,21 g/mol Kokepunkt: 170°C Smeltepunkt: 36 °C Tetthet: 1,44 g/cm³ (20 °C) |
MDL nummer:
MFCD00007126 CAS-nummer: 86-00-0 EINECS: 201-646-8 Merck Indeks: 14,06591 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...