6-Chlorochroman-4-one ≥98.0% (by GC)

Leverandør: TCI

C1968-25G C1968-5G
TCIAC1968-25GEA 4875 NOK
TCIAC1968-25G TCIAC1968-5G
6-Chlorochroman-4-one ≥98.0% (by GC)
6-Chlorochroman-4-one
Formel: C₉H₇ClO₂
MW: 182,61 g/mol
MDL nummer: MFCD00024070
CAS-nummer: 37674-72-9
EINECS: 253-611-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21c1806a7309bfd87ebcf0ace8083c791725af41&vid=21c1806a7309bfd87ebcf0ace8083c791725af41&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR