2-Amino-9-fluorenone ≥98.0% (by HPLC)
Leverandør: TCI
Warning
Synonymer: 2-Amino-9H-fluoren-9-one
A1040-1G A1040-5G
TCIAA1040-1GEA 540 NOK
TCIAA1040-1G TCIAA1040-5G
2-Amino-9-fluorenone ≥98.0% (by HPLC)
2-Amino-9-fluorenone
Formel: C₁₃H₉NO
MW: 195,22 g/mol
Kokepunkt: 361 °C (1013 hPa)
Smeltepunkt: 163 °C
MDL nummer: MFCD00001157
CAS-nummer: 3096-57-9
EINECS: 221-445-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39352b28259ca900b5edefae93ff31ec41864e44&vid=39352b28259ca900b5edefae93ff31ec41864e44&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 156.0 to 162.0 deg-C
Solubility in hot Methanol almost transparency