6-Chloronicotinic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid
C1116-25G C1116-250G C1116-100G
TCIAC1116-25GEA 970 NOK
TCIAC1116-25G TCIAC1116-250G TCIAC1116-100G
6-Chloronicotinic acid ≥98.0% (by titrimetric analysis)
6-Chloronicotinic acid
Formel: C₆H₄ClNO₂
MW: 157,56 g/mol
Kokepunkt: 284 °C (1013 hPa)
Smeltepunkt: 188…189 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006241
CAS-nummer: 5326-23-8
EINECS: 226-201-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c362a62664f042a2af724498af595a35f97d007a&vid=c362a62664f042a2af724498af595a35f97d007a&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %