2-Phenylthiophene ≥95.0% (by GC)
Leverandør: TCI
P1397-1G
P1397-5G
TCIAP1397-1GEA
340
NOK
TCIAP1397-1G
TCIAP1397-5G
2-Phenylthiophene ≥95.0% (by GC)
2-Phenylthiophene
Formel:
C₁₀H₈S MW: 160,24 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00130080 CAS-nummer: 825-55-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 34.0 to 38.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...