(S)-(+)-3-Chloro-1,2-propanediol ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
(2S)-(+)-3-Chloropropane-1,2-diol, (S)-(+)-3-Chloro-1,2-dihydroxy propane
C1542-10G
C1542-25G
TCIAC1542-10GEA
525
NOK
TCIAC1542-10G
TCIAC1542-25G
(S)-(+)-3-Chloro-1,2-propanediol ≥98.0% (by GC)
(S)-(+)-3-Chloro-1,2-propanediol
Formel:
ClCH₂CHOHCH₂OH MW: 110,54 g/mol Kokepunkt: 80°C Tetthet: 1,322 g/cm³ (20 °C) Flash punkt: 126°C |
MDL nummer:
MFCD00210270 CAS-nummer: 60827-45-4 FN: 2689 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...