tert-Butyl acetoacetate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: tert-Butyl Acetoacetate

A0816-25ML A0816-500ML A0816-100ML
TCIAA0816-25MLEA 220 NOK
TCIAA0816-25ML TCIAA0816-500ML TCIAA0816-100ML
tert-Butyl acetoacetate ≥95.0% (by GC)
tert-Butyl acetoacetate
Formel: C₈H₁₄O₃
MW: 158,2 g/mol
Kokepunkt: 184°C
Smeltepunkt: –38 °C
Tetthet: 0,9756 g/cm³ (20 °C)
Flash punkt: 66°C
MDL nummer: MFCD00008811
CAS-nummer: 1694-31-1
EINECS: 216-904-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1468c14f572b5ca843900e594abe7bb3e6048051&vid=1468c14f572b5ca843900e594abe7bb3e6048051&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) 95.0% (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR