Methyl acetoacetate ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Methyl 3-oxobutanoate
A0650-25G
A0650-500G
TCIAA0650-25GEA
220
NOK
TCIAA0650-25G
TCIAA0650-500G
Methyl acetoacetate ≥99.0% (by GC)
Methyl acetoacetate
Formel:
C₅H₈O₃ MW: 116,12 g/mol Kokepunkt: 170°C Smeltepunkt: –80°C Tetthet: 1,076 g/cm³ (20 °C) Flash punkt: 72°C |
MDL nummer:
MFCD00008784 CAS-nummer: 105-45-3 EINECS: 203-299-8 Merck Indeks: 14,06009 |
Spesifikasjon Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |