Ethyl undecanoate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Undecylic acid ethyl ester, Undecanoic acid ethyl ester

U0049-25ML U0049-250ML
TCIAU0049-25MLEA 300 NOK
TCIAU0049-25ML TCIAU0049-250ML
Ethyl undecanoate ≥98.0% (by GC)
Ethyl undecanoate
Formel: C₁₃H₂₆O₂
MW: 214,35 g/mol
Kokepunkt: 140°C
Smeltepunkt: –14,7 °C
Tetthet: 0,86 g/cm³ (20 °C)
MDL nummer: MFCD00008958
CAS-nummer: 627-90-7
EINECS: 211-018-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=922d4630142979b79c477abfbc8ff6bcf06c234b&vid=922d4630142979b79c477abfbc8ff6bcf06c234b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR