Butyl nitrite ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Nitrous acid butyl ester

N0376-25ML N0376-500ML
TCIAN0376-25MLEA 380 NOK
TCIAN0376-25ML TCIAN0376-500ML
Butyl nitrite ≥95.0% (by GC)
Butyl nitrite
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Kokepunkt: 76°C
Tetthet: 0,88 g/cm³ (20 °C)
Flash punkt: 10 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00002058
CAS-nummer: 544-16-1
EINECS: 208-862-1
FN: 2351
ADR: 3,II
Merck Indeks: 14,01582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a8e31b85e2c2c2ff06044b8e94e3a2d6f6b64a8&vid=1a8e31b85e2c2c2ff06044b8e94e3a2d6f6b64a8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR