Butyl isobutyrate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Isobutyric acid butyl ester
I0315-25ML
I0315-500ML
TCIAI0315-25MLEA
220
NOK
TCIAI0315-25ML
TCIAI0315-500ML
Butyl isobutyrate ≥98.0% (by GC)
Butyl isobutyrate
Formel:
C₈H₁₆O₂ MW: 144,21 g/mol Kokepunkt: 150°C Flash punkt: 43°C |
MDL nummer:
MFCD00048773 CAS-nummer: 97-87-0 EINECS: 202-614-6 FN: 3272 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...