2-Klor-4-metokysfenol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Chlor-4-hydroxyanisole

C1487-25G C1487-5G
TCIAC1487-25GEA 6025 NOK
TCIAC1487-25G TCIAC1487-5G
2-Klor-4-metokysfenol ≥98.0% (by GC)
2-Klor-4-metokysfenol
Formel: C₇H₇ClO₂
MW: 158,58 g/mol
Kokepunkt: 242 °C (1013 hPa)
Smeltepunkt: 46…47 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00070773
CAS-nummer: 18113-03-6
EINECS: 242-007-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71d85cefb00701fd9ccf27675a5edf86fc67d06b&vid=71d85cefb00701fd9ccf27675a5edf86fc67d06b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR