4-Iodo-o-toluidine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 2-Amino-5-iodotoluene, 4-Iodo-2-methylaniline
I0690-25G I0690-5G
TCIAI0690-25GEA 1225 NOK
TCIAI0690-25G TCIAI0690-5G
4-Iodo-o-toluidine ≥98.0% (by GC, titration analysis)
4-Iodo-o-toluidine
Formel: C₇H₈IN
MW: 233,05 g/mol
Kokepunkt: 279 °C (1013 hPa)
Smeltepunkt: 91…92 °C
MDL nummer: MFCD00025299
CAS-nummer: 13194-68-8
EINECS: 236-154-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f37e83a3e924269f9df1a93ad74ac68f63b6b501&vid=f37e83a3e924269f9df1a93ad74ac68f63b6b501&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Purple to Dark purple to Dark red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 85.0 to 91.0 deg-C