4'-Isopropylacetophenone ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
p-Isopropylacetophenone, 4-Isopropylacetophenone
I0570-25ML
I0570-5ML
TCIAI0570-25MLEA
2200
NOK
TCIAI0570-25ML
TCIAI0570-5ML
4'-Isopropylacetophenone ≥95.0% (by GC)
4'-Isopropylacetophenone
Formel:
C₁₁H₁₄O MW: 162,23 g/mol Kokepunkt: 238°C Tetthet: 0,97 g/cm³ (20 °C) |
MDL nummer:
MFCD00048297 CAS-nummer: 645-13-6 EINECS: 211-433-1 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...