4-Methoxybiphenyl ≥99.0% (by GC)
Leverandør: TCI
Synonymer:
Biphenyl-4-yl methyl ether, 4-Phenylanisole
M0944-25G
M0944-5G
TCIAM0944-25GEA
770
NOK
TCIAM0944-25G
TCIAM0944-5G
4-Methoxybiphenyl ≥99.0% (by GC)
4-Methoxybiphenyl
Formel:
C₁₃H₁₂O MW: 184,24 g/mol Kokepunkt: 157°C |
MDL nummer:
MFCD00014897 CAS-nummer: 613-37-6 EINECS: 210-339-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 87.0 to 90.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...