1-Acetylnaphthalene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

A0052-25G A0052-500G
TCIAA0052-25GEA 395 NOK
TCIAA0052-25G TCIAA0052-500G
1-Acetylnaphthalene ≥98.0% (by GC)
1-Acetylnaphthalene
Formel: C₁₂H₁₀O
MW: 170,21 g/mol
Kokepunkt: 298°C
Smeltepunkt: 9…11 °C
Tetthet: 1,12 g/cm³ (20 °C)
Flash punkt: ≥ 100 °C
MDL nummer: MFCD00004013
CAS-nummer: 941-98-0
EINECS: 213-384-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69317198d2e8c67b2c4e4ecd7023a0a517cf2182&vid=69317198d2e8c67b2c4e4ecd7023a0a517cf2182&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR