2-Aminopyrazine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Aminopyrazine, Pyrazin-2-amine

A0989-25G A0989-5G
TCIAA0989-25GEA 875 NOK
TCIAA0989-25G TCIAA0989-5G
2-Aminopyrazine ≥98.0% (by GC, titration analysis)
2-Aminopyrazine
Formel: C₄H₅N₃
MW: 95,1 g/mol
Kokepunkt: 203 °C (1013 hPa)
Smeltepunkt: 117…120 °C
MDL nummer: MFCD00006137
CAS-nummer: 5049-61-6
EINECS: 225-748-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c0445f13bdae689ec317c45b17175d96cead041&vid=7c0445f13bdae689ec317c45b17175d96cead041&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR