Cyanuric acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

C0459-25G C0459-500G
TCIAC0459-25GEA 220 NOK
TCIAC0459-25G TCIAC0459-500G
Cyanuric acid ≥98.0% (by GC, titration analysis)
Cyanuric acid
Formel: C₃H₃N₃O₃
MW: 129,08 g/mol
Tetthet: 2,5 g/cm³ (20 °C)
MDL nummer: MFCD00082990
CAS-nummer: 108-80-5
EINECS: 203-618-0
Merck Indeks: 14,02698
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee52dc82eb9ccc79d7b43dab1c6c7718abb8f78f&vid=ee52dc82eb9ccc79d7b43dab1c6c7718abb8f78f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Sodium hydroxide solution almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR