Benzoylacetonitrile ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Phenyl-3-oxopropanenitrile, 3-Oxo-3-phenylpropanenitrile

B0098-25G B0098-5G
TCIAB0098-25GEA 985 NOK
TCIAB0098-25G TCIAB0098-5G
Benzoylacetonitrile ≥98.0% (by GC)
Benzoylacetonitrile
Formel: (C₆H₅)C(O)CH₂CN
MW: 145,16 g/mol
Kokepunkt: 160°C
Smeltepunkt: 83,5 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00001942
CAS-nummer: 614-16-4
EINECS: 210-369-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dc52444bbc4b25f11f30f5d96c253b8d5e87bb2&vid=1dc52444bbc4b25f11f30f5d96c253b8d5e87bb2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR