N-(3-Bromopropyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
B1328-25G
B1328-500G
TCIAB1328-25GEA
675
NOK
TCIAB1328-25G
TCIAB1328-500G
N-(3-Bromopropyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(3-Bromopropyl)phthalimide
Formel:
C₁₁H₁₀BrNO₂ MW: 268,11 g/mol Kokepunkt: 433 °C (1013 hPa) Smeltepunkt: 70…73 °C |
MDL nummer:
MFCD00005904 CAS-nummer: 5460-29-7 EINECS: 226-738-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 72.0 to 75.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...