N-(4-Bromobutyl)phthalimide ≥95.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-(4-Bromobutyl)isoindoline-1,3-dione

B1394-25G B1394-5G
TCIAB1394-25GEA 1700 NOK
TCIAB1394-25G TCIAB1394-5G
N-(4-Bromobutyl)phthalimide ≥95.0% (by HPLC, titration analysis)
N-(4-Bromobutyl)phthalimide
Formel: C₁₂H₁₂BrNO₂
MW: 282,14 g/mol
Smeltepunkt: 77…80 °C
MDL nummer: MFCD00005905
CAS-nummer: 5394-18-3
EINECS: 226-401-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49116b06eac74063a015fa89f77bd3701ba806ed&vid=49116b06eac74063a015fa89f77bd3701ba806ed&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR