4-(Trifluoromethyl)benzylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

T1508-25G T1508-5G
TCIAT1508-25GEA 1875 NOK
TCIAT1508-25G TCIAT1508-5G
4-(Trifluoromethyl)benzylamine ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)benzylamine
Formel: C₈H₈F₃N
MW: 175,15 g/mol
Kokepunkt: 113°C
Tetthet: 1,229 g/cm³ (20 °C)
Flash punkt: 75°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00010220
CAS-nummer: 3300-51-4
EINECS: 221-971-9
FN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b62a53cd20aa338ac5d8c181948e9fabb1cbe97&vid=4b62a53cd20aa338ac5d8c181948e9fabb1cbe97&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR