2-Nitro-4-(trifluoromethoxy)aniline ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
N0841-25G
N0841-5G
TCIAN0841-25GEA
1050
NOK
TCIAN0841-25G
TCIAN0841-5G
2-Nitro-4-(trifluoromethoxy)aniline ≥98.0% (by GC)
2-Nitro-4-(trifluoromethoxy)aniline
Formel:
C₇H₅F₃N₂O₃ MW: 222,12 g/mol |
MDL nummer:
MFCD00042326 CAS-nummer: 2267-23-4 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...