3-Fluoro-p-anisidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Fluoro-4-anisidine, 3-Fluoro-4-methoxyaniline, 4-Amino-2-fluoroanisole

F0648-25G F0648-5G
TCIAF0648-25GEA 2600 NOK
TCIAF0648-25G TCIAF0648-5G
3-Fluoro-p-anisidine ≥98.0% (by GC, titration analysis)
3-Fluoro-p-anisidine
Formel: C₇H₈FNO
MW: 141,15 g/mol
Kokepunkt: 135°C
MDL nummer: MFCD00075040
CAS-nummer: 366-99-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95cdf8fb9bbe4aa2ec3580911b0c8c2166477a02&vid=95cdf8fb9bbe4aa2ec3580911b0c8c2166477a02&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR