4-Bromo-2-chloro-1-iodobenzene ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Bromo-3-chloro-4-iodobenzene

B3030-25G B3030-5G
TCIAB3030-25GEA 1925 NOK
TCIAB3030-25G TCIAB3030-5G
4-Bromo-2-chloro-1-iodobenzene ≥97.0% (by GC)
4-Bromo-2-chloro-1-iodobenzene
Formel: C₆H₃BrClI
MW: 317,35 g/mol
Kokepunkt: 104°C
MDL nummer: MFCD00079709
CAS-nummer: 31928-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df3a8e9dadd560b5acb5c515254c696832351d30&vid=df3a8e9dadd560b5acb5c515254c696832351d30&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR