5-Fluoro-o-toluonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

F0747-25G F0747-5G
TCIAF0747-25GEA 1875 NOK
TCIAF0747-25G TCIAF0747-5G
5-Fluoro-o-toluonitrile ≥98.0% (by GC)
5-Fluoro-o-toluonitrile
Formel: C₈H₆FN
MW: 135,14 g/mol
Kokepunkt: 226 °C (1013 hPa)
Smeltepunkt: 42…46 °C
MDL nummer: MFCD00042295
CAS-nummer: 77532-79-7
FN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c75698f6284c32338f77fbe5f5368292bc1ba2d5&vid=c75698f6284c32338f77fbe5f5368292bc1ba2d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR