Furfuryl mercaptan ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-Furanmethanethiol, 2-Furanmethanthiol, 2-Furylmethanethiol

F0077-25G F0077-500G
TCIAF0077-25GEA 380 NOK
TCIAF0077-25G TCIAF0077-500G
Furfuryl mercaptan ≥98.0% (by GC)
Furfuryl mercaptan
Formel: C₅H₆OS
MW: 114,17 g/mol
Kokepunkt: 155°C
Tetthet: 1,1339 g/cm³ (20 °C)
Flash punkt: 45°C
MDL nummer: MFCD00003254
CAS-nummer: 98-02-2
EINECS: 202-628-2
FN: 1228
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f79d1db0282025769a1b5fefc8a8e3234751b31d&vid=f79d1db0282025769a1b5fefc8a8e3234751b31d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR