2,2,3,3-Tetrafluoro-1,4-butanediol ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
2,2,3,3-Tetrafluorobutane-1,4-diol
T2296-1G
T2296-5G
T2296-25G
TCIAT2296-1GEA
285
NOK
TCIAT2296-1G
TCIAT2296-5G
TCIAT2296-25G
2,2,3,3-Tetrafluoro-1,4-butanediol ≥95.0% (by GC)
2,2,3,3-Tetrafluoro-1,4-butanediol
Formel:
C₄H₆F₄O₂ MW: 162,08 g/mol Kokepunkt: 112°C Smeltepunkt: 85 °C |
MDL nummer:
MFCD00042375 CAS-nummer: 425-61-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...